Learn More
Cayman Chemical MultIubIquItIn ChaIn MoncLN
SDP
Supplier:  Cayman Chemical 14220100 UG
Encompass_Preferred
Immunogen: Crude preparation of polyubiquitinated-lysozyme • Host: Mouse, Clone FK2 • Cross Reactivity: (+) Mono- and polyubiquitinated conjugates; (−) Free ubiquitin • Application(s): ELISA, IHC, WB
Catalog No. 50-295-4519