Learn More
Medchemexpress LLC mUNO | 2283322-63-2 | 97.5% | 561.65 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP10894100MG
Encompass_Preferred
mUNO is a tumor-homing peptide with the sequence "CSPGAK" that specifically binds to mouse CD206, which targets tumor-associated macrophages expressing CD206/MRC1. It can also interact with human recombinant CD206. In vitro studies showed that FAM-mUNO (3 nM; 45 min) selectively binds to IL-4-differentiated CD206+ human monocyte-derived macrophages. It is intended for research use only.
  • Specifically binds to mouse CD206
  • Targets tumor-associated macrophages expressing CD206/MRC1
  • Interacts with human recombinant CD206
  • Selectively binds to IL-4-differentiated CD206+ human monocyte-derived macrophages
  • Intended for research use
Catalog No. 50-004-23076