Learn More
Medchemexpress LLC Muno | 2283322-63-2 | 97.5% | 561.65 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP1089450MG
Encompass_Preferred
mUNO is a tumor-homing peptide (sequence: "CSPGAK") that specifically binds to mouse CD206, targeting tumor-associated macrophages that express CD206/MRC1. It can also interact with human recombinant CD206.
  • Specifically binds to mouse CD206
  • Targets tumor-associated macrophages
  • Interacts with human recombinant CD206
Catalog No. 50-004-22391