Learn More
Medchemexpress LLC Mupirocin calcium hydrate | 115074-43-6 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYN706810MM/1ML
Encompass_Preferred
Mupirocin calcium hydrate is an orally active antibiotic isolated from *Pseudomonas fluorescens*. It inhibits bacterial protein and RNA synthesis by reversibly inhibiting isoleucyl-transfer RNA. This product is for research use only.
  • Exhibits antibacterial effect against staphylococci, streptococci and certain gram-negative bacteria
  • Decreases total bacterial loads in skin lesions
  • Alleviates MRSA-infected pressure ulcers
  • Prevents vascular prosthetic graft infection due to *Staphylococcus epidermidis*
Catalog No. 50-004-19683