Learn More
Medchemexpress LLC Mupirocin calcium hydrate | 115074-43-6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN706850MG
Encompass_Preferred
Mupirocin calcium hydrate is an orally active antibiotic isolated from *Pseudomonas fluorescens*. It exerts its antimicrobial activity by reversibly inhibiting isoleucyl-transfer RNA, thereby inhibiting bacterial protein and RNA synthesis.
  • Antibacterial agent
  • Orally active
  • Inhibits isoleucyl-transfer RNA
  • Inhibits bacterial protein and RNA synthesis
  • Purity: 98.05%
Catalog No. 50-004-18954