Learn More
U.S. Pharmacopeia Mupirocin Lithium, 73346-79-9, MFCD11977847, 100mg
Supplier:  U.S. Pharmacopeia 1448923
Encompass
Molecular formula C26H44O9 . Li, Molecular weight 506.56, Synonyms: Lithium mupirocin, Lithium pseudomonate
Catalog No. NC9022090