Learn More
Cayman Chemical Muramyl DIpeptIde 5mg
SDP
Supplier:  Cayman Chemical 308665 MG
Encompass_Preferred
A NOD2 ligand; binds to NOD2 to induce NF-κB signaling and the production of pro-inflammatory cytokines, chemokines, and antimicrobial peptides; has been used as an adjuvant to enhance non-specific immunity to bacterial infections in mice
Catalog No. 50-297-8843