Learn More
Alkali Scientific Murashige and Skoog Medium with Vitamins, Van der Salm Modification, 22 Grams of Powder, Makes 5 Liters of Solution
SDP
Supplier:  Alkali Scientific KOO003CP
Encompass_Preferred
Murashige and Skoog Medium with Vitamins (Van der Salm modification) is a powder medium designed for plant tissue culture applications. This formula, which makes 5 liters of solution from 22 grams of powder, is used to promote the growth of plant cells and tissues under controlled laboratory conditions. The Van der Salm modification ensures optimized plant growth and is particularly useful for shoot regeneration and plant propagation experiments. This medium is typically used in experiments where precise growth conditions are needed for various plant species. It supports the development of healthy plants in vitro by providing essential nutrients and growth factors, including vitamins that support cellular metabolism.
Catalog No. 50-006-59921