Learn More
Alkali Scientific Murashige and Skoog Modified Medium, 5 L
SDP
Supplier:  Alkali Scientific IGE004CP
Encompass_Preferred
Murashige and Skoog Modified Medium (MS), 5 L, is designed for plant tissue culture applications, providing a stable environment for the growth of plant cells and tissues. This modified version of the Murashige and Skoog (MS) medium offers the necessary nutrients, vitamins, and hormones to promote plant growth and development in a controlled in vitro setting. Ideal for research labs and small-scale tissue culture operations, the 5 L quantity ensures ample supply for multiple experiments. This medium is used in plant regeneration, callus formation, and studies on plant growth factors, making it essential for plant biotechnologists and researchers working with various species.
Catalog No. 50-006-59677