Learn More
Bioworld Murashige and Skoog (MS) Medium, Buffered, 25 L
Supplier:  Bioworld 306300643
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Macronutrients and Micronutrients as described by Murashige & Skoog (1967)Buffered with MES MES is an excellent buffer for plant media because of high buffer capacity and its pH range of 5.5-6.7 Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Working Concentration: 4.83 g/LProvided as 1 unit to make 10L, 25L, or 50L of mediumPlant tissue culture tested
Catalog No. 50-254-679