Learn More
Bioworld Murashige and Skoog (MS) Medium w/ Vitamins, 25 L
Supplier:  Bioworld 306300672
Encompass_Preferred
Murashige & Skoog (MS) MediumContains the Macronutrients, Micronutrients, and Vitamins as described by Murashige & Skoog (1962)Cytokines, Sucrose, and Auxins can be added as needed Sold in individual containers with enough medium to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 4.43 g/LpH: 6−7Soluble in water
Catalog No. 50-254-688