Learn More
Medchemexpress LLC HY-P1674A 5mg Medchemexpress, Murepavadin (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1674A5MG
Encompass_Preferred
Medchemexpress, HY-P1674A 5mg Murepavadin (TFA) CAS: Murepavadin (TFA), a 14-amino-acid cyclic peptide, is a highly potent, specific antibiotic for the treatment of bacterial infections caused by Pseudomonas aeruginosa. Murepavadin (TFA) targets the lipopolysaccharide transport portin D [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4238