Learn More
Medchemexpress LLC Murexide (ammonium purpurate) | 3051-09-0 | MFCD00012777 | 98.0% | 284.19 g·mol⁻¹ | C8H8N6O6 | 50G
SDP
Supplier:  Medchemexpress LLC HY12592950G
Encompass_Preferred
Murexide (ammonium purpurate) is a purple solid reagent used as a colorimetric indicator in analytical chemistry. It is commonly used in complexometric titrations to determine water hardness and to detect certain metal ions. The material is supplied as a solid powder with typical purity of 98% and has defined storage recommendations for powder and in-solution stability.
Catalog No. 50-002-36515