Learn More
Cayman Chemical MurIdeoxycholIc AcId 10mg
SDP
Supplier:  Cayman Chemical 2029010 MG
Encompass_Preferred
A secondary bile acid produced from α-muricholic acid and β-muricholic acid; metabolite of lithocholic acid; prevents gallstones in hamsters; does not resolve gallstones in prairie dogs and induces formation of insoluble bile acid salt gallstones; rapidly absorbed in humans
Catalog No. 50-298-1714