Learn More
ATCC Murine adenovirus 1; Strain: FL
Supplier:  ATCC VR550
Encompass
Murine adenovirus 1 strain FL is propagated in BALB/3T3 clone A31 cells (ATCC CCL-163). This strain was isolated from the spleen pool of mice infected with the Friend leukemia virus. The strain has applications in immune system disorder research.
Catalog No. 50-238-4282