Learn More
Eagle Biosciences Inc Recombinant murine leukemia virus p30 protein, 10 ug
SDP
Supplier:  Eagle Biosciences Inc P500100
Encompass
Gag polyprotein plays a role in budding and is processed by the viral protease during virion maturation outside the cell. Capsid protein p30 forms the spherical core of the virion that encapsulates the genomic RNA-nucleocapsid complex.
Catalog No. 50-187-8419