Learn More
ATCC Murine pneumonia virus; Strain: 15; Pneumonia virus of mice
Supplier:  ATCC VR1819
Encompass
Murine pneumonia virus strain 15 is propagated in BHK-21 [C-13] cells (ATCC CCL-10). This animal virus was isolated from the lungs of mice and was treated for Mycoplasma sp. using a mycoplasma removal agent.
Catalog No. 50-238-5062