Learn More
Cayman Chemical MurIsteron A 1mg
SDP
Supplier:  Cayman Chemical 117411 MG
Encompass_Preferred
A natural analog of 20-hydroxyecdysone and agonist of the ecdysteroid receptor (Kd = 1 nM); used as an agonist in ecdysone-inducible gene expression systems in mammalian cells
Catalog No. 50-295-5951