Learn More
Medchemexpress LLC Muscone | 541-91-3 | 238.41 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0633100MG
Encompass_Preferred
Muscone is the primary active monomer derived from traditional Chinese medicine musk. It functions by inhibiting NF-κB and NLRP3 inflammasome activation. This action leads to a significant reduction in the levels of inflammatory cytokines such as IL-1β, TNF-α, and IL-6, ultimately enhancing cardiac function and improving the survival rate.
  • Main active monomer of traditional Chinese medicine musk.
  • Inhibits NF-κB and NLRP3 inflammasome activation.
  • Decreases inflammatory cytokines (IL-1β, TNF-α, and IL-6).
  • Improves cardiac function and survival rate.
Catalog No. 50-003-88360