Learn More
Medchemexpress LLC Muscone | 541-91-3 | 238.41 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN063350MG
Encompass_Preferred
Muscone is the primary active monomer derived from traditional Chinese medicine musk. It functions by inhibiting the activation of NF-κB and NLRP3 inflammasome. This action leads to a significant reduction in inflammatory cytokines such as IL-1β, TNF-α, and IL-6, ultimately enhancing cardiac function and improving survival rates.
  • Functions by inhibiting the activation of NF-κB and NLRP3 inflammasome.
  • Leads to a significant reduction in inflammatory cytokines such as IL-1β, TNF-α, and IL-6.
  • Enhances cardiac function.
  • Improves survival rates.
Catalog No. 50-003-88691