Learn More
Medchemexpress LLC Mussaenosidic acid | 82451-22-7 | 376.36 | 5 MG

Supplier: Medchemexpress LLC HYN125805MG
Mussaenosidic acid is an active product extracted from Pedicularis kerneri Dalla Torre. It exhibits weak antiglycation activity.
- Purity: 98.84%
- Appearance: Solid
- Color: White to off-white
- Initial source: Plants, Orobanchaceae, Pedicularis kerneri Dalla Torre
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.