Learn More
Selleck Chemical LLC MV1035-E1636-25MG
SDP
Supplier:  Selleck Chemical LLC E163625MG
Encompass_Preferred
MV1035 an inhibitor of ALKBH5 significantly reduces GBM U87-MG cell migration and invasiveness through inhibition of the RNA demethylase ALKBH5
Catalog No. 50-313-6586