Learn More
ABclonal Technology MVB12A Rabbit pAb
SDP
Supplier:  ABclonal Technology A15933500UL
Encompass_Preferred
Enables lipid binding activity and ubiquitin binding activity. Involved in regulation of epidermal growth factor receptor signaling pathway, viral budding, and virus maturation. Located in several cellular components, including Golgi apparatus, centrosome, and nucleoplasm. Part of ESCRT I complex.
Catalog No. 50-001-04492