Learn More
MVE BIOLOGICAL SOLUTIONS MVE CryoShipper 2000
Supplier:  MVE BIOLOGICAL SOLUTIONS 11366673
Encompass_Preferred
The MVE CryoShipper 2000 is engineered specifically to ship large quantities of 1.2 and 2.0 mL vials. The new design can also store samples in liquid nitrogen after arrival at its destination.
Catalog No. NC1331828