Learn More
Medchemexpress LLC My-673 | 2944459-96-3 | 98.6% | 322.31 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14941410MM/1ML
Encompass_Preferred
MY-673 is a colchicine binding site inhibitor (CBSI) that effectively inhibits tubulin polymerization. This compound also targets and inhibits the ERK signaling pathway, which in turn influences the expression levels of SMAD4 protein within the TGF-β/SMAD pathway. Studies have shown that MY-673 can inhibit cell proliferation and migration while inducing apoptosis both in vivo and in vitro.
  • Inhibits tubulin polymerization
  • Inhibits ERK signaling pathway
  • Influences SMAD4 protein expression
  • Inhibits cell proliferation and migration
  • Induces apoptosis
Catalog No. 50-003-60838