Learn More
Medchemexpress LLC My-875 | 2974348-62-2 | MFCD34676541 | 99.6% | 387.43 | C21H25NO6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15076110MG
Encompass_Preferred
MY-875 is a competitive microtubulin polymerization inhibitor that targets the colchicine binding site and activates the Hippo signaling pathway. It induces apoptosis, causes G2/M cell-cycle arrest, and exhibits dose-dependent antiproliferative activity in multiple cancer cell lines; reported tubulin polymerization IC50 is 0.92 μM. The compound is supplied as a high-purity solid with high solubility in DMSO and recommended storage to preserve stability.
  • Competitive microtubulin polymerization inhibitor targeting colchicine binding sites.
  • Activates Hippo pathway leading to YAP degradation.
  • Induces apoptosis and G2/M cell-cycle arrest in cancer cells.
  • Demonstrates potent antiproliferative activity (tubulin polymerization IC50 ~0.92 μM).
  • High purity solid with high DMSO solubility for assay preparation.
  • Stable under recommended storage conditions for experimental reproducibility.
Catalog No. 50-002-87157