Learn More
Medchemexpress LLC MY-875 | C21H25NO6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15076110MM/1ML
Encompass_Preferred
MY-875 is a competitive inhibitor of microtubulin polymerization with an IC50 value of 0.92 μM. It works by targeting colchicine binding sites and activating the Hippo pathway. This compound induces apoptosis and demonstrates anticancer activity. It is intended for research use only.
  • Inhibits microtubulin polymerization
  • Targets colchicine binding sites
  • Activates the Hippo pathway
  • Induces apoptosis
  • Exhibits anticancer activity
  • Shows significant anti-proliferative activity against cancer cells
  • Inhibits cell colony-forming ability
  • Arrests cells in the G2/M phase
  • Reduces expression of anti-apoptotic proteins
Catalog No. 50-003-75465