Learn More
Sigma Aldrich Fine Chemicals Biosciences MyBlockTM Mini dry bath wi1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences BMSBSH3001EA
Encompass
Capable of heating as well as cooling test samplesMinimum Programmable temp. 0 CCompact fits in the palm of your handExchangeable blocks for 0.2 to 50ml tubes
Catalog No. 50-290-8721