Learn More
Medchemexpress LLC MYC-IN-3 | 3042878-82-7 | 98.7% | 543.89 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY162686100MG
Encompass_Preferred
MYC-IN-3 (compound 37) is a potent MYC inhibitor that demonstrates antiproliferative and anticancer activity. It shows potential for research related to prostate cancer.
  • Potent MYC inhibitor.
  • Shows antiproliferative activity.
  • Shows anticancer activity.
  • Has potential for the research of prostate cancer.
Catalog No. 50-003-92771