Learn More
Proteintech Group Inc Myc-Trap Agarose
SDP
Supplier:  Proteintech Group Inc YTA20
Encompass
Myc-Trap Agarose is an affinity reagent for IP and purification of Myc-tagged proteins. It consists of a Myc Nanobody/ VHH coupled to agarose beads.
Catalog No. 50-225-8937
 Restricted Item