Learn More
Cayman Chemical Myca uba bak Myctn 100mg
SDP
Supplier:  Cayman Chemical 26902100 MG
Encompass_Preferred
A flavonoid with diverse biological activities; active against B. subtilis, E. coli, K. pneumoniae, S. aureus, and C. albicans (MIC = 0.25 mg/ml for all); inhibits AChE and α-glucosidase in vitro (IC50s = 65 and 79 μg/ml, respectively); inhibits LPS-induced production of TNF-α and enhances LPS-induced RANTES production in RAW 264.7 cells at 0.2 and 1 mM
Catalog No. 50-294-1470