Learn More
Medchemexpress LLC HY-129600 10mg , MYCi361 CAS:2289690-31-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12960010MG
Encompass_Preferred
HY-129600 10mg MYCi361 CAS: 2289690-31-7 MYCi361 (NUCC-0196361) is a MYC inhibitor with the Kd of 3.2 μM for binding to MYC. MYCi361 (NUCC-0196361) suppresses tumor growth and enhances anti-PD1 immunotherapy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0026