Learn More
Cayman Chemical MyclobutnIl-d9 5mg
SDP
Supplier:  Cayman Chemical 392635 MG
Encompass_Preferred
An internal standard for the quantification of myclobutanil by GC- or LC-MS
Catalog No. 50-291-8513