Learn More
Apexbio Technology LLC MYCMI-6 681282-09-7 50mg
SDP
Supplier:  Apexbio Technology LLC BA276650
Encompass_Preferred
MYCMI-6 (CAS 681282-09-7) is a selective inhibitor of endogenous protein-protein interactions specifically targeting the basic helix loop helix leucine zipper (bHLHZip) domain of MYC with a binding affinity of 1 6 M By disrupting interactions at this domain MYCMI-6 blocks MYC-driven transcriptional activity The compound inhibits the proliferation of tumor cells in a MYC-dependent manner with IC50 values below 0 5 M while displaying no cytotoxicity toward normal human cells MYCMI-6 also induces apoptosis in treated cells It is utilized in research settings to investigate MYC function and the consequences of MYC inhibition in cancer biology
Catalog No. 50-000-89504