Learn More
ATCC Mycobacterium chelonae
Supplier:  ATCC 35752
Encompass
Mycobacterium chelonae strain TMC 1544 [Friedmann] is a whole-genome sequenced type strain that was isolated from a tortoise. This bacterial strain has applications in sterility assurance and testing an antimicrobial agents.
Catalog No. 50-238-3991