Learn More
ATCC Alcanivorax borkumensis
Supplier:  ATCC 700651
Encompass
Alcanivorax borkumensis is a whole-genome sequenced bacterial type strain that was isolated in 1991 from sea water sediment in the Isle of Borkum. This strain is cited to degrade n-alkanes and to produce glucose lipids.
Catalog No. 50-238-1448