Learn More
ATCC Mycobacterium tuberculosis;Strain H37Rv; genomic DNA
Supplier:  ATCC 25618D2
Encompass
Genomic DNA from Mycobacterium tuberculosis strain H37Rv (ATCC 25618). This product can be used in PCR or other molecular biology applications.
Catalog No. 50-238-3995