Learn More
Selleck Chemical LLC Mycophenolate Mofetil 50mg 128794-94-5 RS61443
SDP
Supplier:  Selleck Chemical LLC S150150MG
Encompass_Preferred
Mycophenolate Mofetil (RS61443) is a non-competitive, selective and reversible inhibitor of inosine monophosphate dehydrogenase I/II with IC50 of 39 nM and 27 nM, respectively. Mycophenolate Mofetil induces caspase-dependent apoptosis and cell cycle inhibition in multiple myeloma cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1325