Learn More
Selleck Chemical LLC Mycophenolic acid 10mM/1mL 24280-93-1 Mycophenolate, RS-61443
SDP
Supplier:  Selleck Chemical LLC S248710MM/1ML
Encompass_Preferred
Mycophenolic acid (Mycophenolate, RS-61443) is a potent IMPDH inhibitor and the active metabolite of an immunosuppressive drug, used to prevent rejection in organ transplantation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2463