Learn More
Medchemexpress LLC Mycophenolic acid | 24280-93-1 | 99.94% | 25 MG

Supplier: Medchemexpress LLC 25MGHYB0421R
Mycophenolic acid is a potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor with an EC50 of 0.24 μM. It demonstrates antiviral effects against a wide range of RNA viruses, including influenza. Mycophenolic acid also acts as an immunosuppressive agent and exhibits antiangiogenic and antitumor effects. It is intended for research use only and not for sale to patients.
- Potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor
- Exhibits antiviral effects against a wide range of RNA viruses, including influenza
- Acts as an immunosuppressive agent
- Shows antiangiogenic and antitumor effects
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.