Learn More
Medchemexpress LLC Mycophenolic acid (sodium) | 37415-62-6 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0421A10MM/1ML
Encompass_Preferred
Mycophenolic acid sodium is a potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor with an EC50 of 0.24 μM. It exhibits antiviral effects against a broad spectrum of RNA viruses and functions as an immunosuppressive agent. This compound also demonstrates antiangiogenic and antitumor properties by modulating the tumor microenvironment. IMPDH is the rate-limiting enzyme in the de novo synthesis of guanosine nucleotides. In vitro, it shows preferential antiproliferative activity against endothelial cells and fibroblasts.
  • Potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor
  • Exhibits antiviral effects against various RNA viruses, including influenza and Zika virus
  • Functions as an immunosuppressive agent
  • Demonstrates antiangiogenic and antitumor properties
  • Modulates tumor microenvironment
  • Causes dose-dependent down-regulation of HDAC2 and MYC
  • Up-regulates NDRG1
Catalog No. 50-004-07893