Learn More
Medchemexpress LLC Mycophenolic acid (sodium) | 37415-62-6 | 99.98% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0421A100MG
Encompass_Preferred
Mycophenolic acid sodium is a potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor. It demonstrates antiviral effects against a wide range of RNA viruses, including influenza, dengue virus, Zika virus, rotavirus, CCHFV, and hantavirus. It is also an immunosuppressive agent and exhibits antiangiogenic and antitumor effects.
  • Potent uncompetitive IMPDH inhibitor
  • Demonstrates antiviral effects against a wide range of RNA viruses
  • Immunosuppressive agent
  • Exhibits antiangiogenic and antitumor effects
  • Preferential antiproliferative activity against endothelial cells and fibroblasts
  • Shows a dose-dependent down-regulation of HDAC2 and MYC
  • Up-regulates NDRG1
Catalog No. 50-004-07892