Learn More
ATCC Mycoplasma arginini; Strain G230
Supplier:  ATCC 23838TTR
Encompass
Mycoplasma arginini strain G230 [NCTC 10129] is a bacterial type strain that was isolated from a mouse brain that was experimentally infected with scrapies. This titered product has applications in assay development, quality control and mycoplasma testing.
Catalog No. 50-238-2849