Learn More
ATCC Mycoplasma Growth Supplement, 10x solution (ATCC 20-2207™)
Supplier:  ATCC 202207
Encompass
Mycoplasma Growth Supplement, 10x solution (ATCC 20-2207™) is a well-defined additive used in broth and agar media to support the growth of various mycoplasma species. Included in ATCC Medium 988 (Spiroplasma SP-4), it contains amino acids, vitamins, nucleotides, and cofactors essential for mycoplasma cultivation.
Catalog No. 50-238-2009