Learn More
ATCC Mycoplasma hominis; Strain PG21
Supplier:  ATCC 23114
Encompass
Mycoplasma hominis strain PG21 [H 50, NCTC 10111] is a whole-genome sequenced bacterial type strain isolated from a rectal swab. This strain produces arginine deiminase.
Catalog No. 50-238-2311