Learn More
ATCC Mycoplasma hominis; Strain PG21; genomic DNA
Supplier:  ATCC 23114D
Encompass
Genomic DNA from Mycoplasma hominis strain PG21 (ATCC 23114). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-2384