Learn More
ATCC Mycoplasma pneumoniae; Strain FH
Supplier:  ATCC 15531TTR
Encompass
Mycoplasma pneumoniae FH strain of Eaton Agent [NCTC 10119] is the first Mycoplasma known to be the etiological agent of a human disease. The strain was isolated by Hayflick from monkey kidney tissue-culture fluids of the FH strain (Eaton Agent Virus) supplied by C. Liu, who recovered this strain in embryonated eggs from a student with atypical pneumonia. This bacterium is provided with titer data (CFU) and calculated genome copy numbers, making it useful for assay development and quality control.
Catalog No. 50-238-1679