Learn More
ATCC Mycoplasma synoviae; Strain WVU 1853
Supplier:  ATCC 25204TTR
Encompass
Mycoplasma synoviae strain WVU 1853 [NCTC 10124] was isolated from the hock joint of a chicken. This titered bacterial type strain can be used in assay development and quality control.
Catalog No. 50-238-3047