Learn More
ATCC Mycoplasma Testing Service: PCR-based
Supplier:  ATCC 136XV
Encompass
FTA Sample Collection Kit for PCR-based Mycoplasma Detection Service combines easy-to-use Whatman FTA sample collection cards with ATCC's best-selling Universal Mycoplasma Detection Kit (ATCC 30-1012K). This kit can quickly and reliably identify over 60 species of Mycoplasma, Acholeplasma, Spiroplasma, and Ureaplasma, including the top eight species most likely to affect cell cultures: M. arginini, M. fermentans, M. hominis, M. hyorhinis, M. orale, M. pirum, M. salivarium, and A. laidlawii. The sample kit is mailed upon purchase. Whatman and FTA are trademarks of the General Electric Company.
Catalog No. 50-238-4025