Learn More
ILEX LIFE SCIENCES LLC MYCORASE 50X SOLUTION 100ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0602100
Encompass_Preferred
PAN-Biotech Mycorase 50X Solution for Mycoplasma Removal (100 ml). DESCRIPTION: Mycorase has been developed to remove a broad range of different strains of mycoplasma in most cell types. Mycorase is easy to use and does not affect eukaryotic cell proliferation. It is a highly effective antibiotic solution for safe eradication of mycoplasma contamination. ADVANTAGES: Ready-to-use solution. Effective removal of mycoplasma. No effect on cell proliferation. Broad range of action. Permanent cure for most cell types. COMPOSITION: Antibiotic solution (50x concentrated) in DPBS for treatment of mycoplasma-infected cell cultures. STORAGE: -20° C (in the dark), thawed at 2-8° C (up to 1 month). MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany.
Catalog No. 50-273-3743